missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrapropylammonium Bisulfate, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Tetrapropylammonium Bisulfate |
| CAS | 56211-70-2 |
| Molecular Formula | C12H29NO4S |
| Quantity | 250 mg |
| Synonym | Tetrapropylammonium Hydrogen Sulfate |
| InChI Formula | InChI=1S/C12H28N.H2O4S/c1-5-9-13(10-6-2,11-7-3)12-8-4;1-5(2,3)4/h5-12H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1 |
| SMILES | CN(C)CCCN1c2c(cc(O)cc2)Sc3c1cc(Cl)cc3 |
| Molecular Weight (g/mol) | 283.43 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction