accessibility menu, dialog, popup

UserName

CAS RN 55620-18-3

CH3OOHOO(E)

CAS RN 55620-18-3

IUPAC Name: (2E)-3-[2-(acetyloxy)phenyl]prop-2-enoic acid
Synonyms: cinnamic acid, 2-acetoxy-
Molecular Weight (g/mol): 206.2
Molecular Formula: C11H10O4
InChi Key: UXOWQQCLBQBRMQ-VOTSOKGWSA-N
SMILES: CC(=O)OC1=CC=CC=C1\C=C\C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results