missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenamine Hydrochloride, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Dibenamine Hydrochloride |
| CAS | 55-43-6 |
| Analyte or Component Names | Dibenamine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H18 Cl N . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benzenemethanamine, N-(2-chloroethyl)-N-(phenylmethyl)-, hydrochloride (1:1), Benzenemethanamine, N-(2-chloroethyl)-N-(phenylmethyl)-, hydrochloride (9CI), Dibenzylamine, N-(2-chloroethyl)-, hydrochloride (8CI), (2-Chloroethyl)dibenzylamine hydrochloride, (N,N-Dibenzylamino)ethyl chloride hydrochloride, 2-(Dibenzylamino)ethyl chloride hydrochloride, Dibenamine hydrochloride, N-(2-Chloroethyl)dibenzylamine hydrochloride, Sympatholytin, (beta-Chloroethyl)dibenzylamine hydrochloride, N,N-Dibenzyl-beta-chloroethylamine hydrochloride |
| InChI Formula | InChI=1S/C16H18ClN.ClH/c17-11-12-18(13-15-7-3-1-4-8-15)14-16-9-5-2-6-10-16;/h1-10H,11-14H2;1H |
| Solution Type | Neat |
| SMILES | Cl.ClCCN(Cc1ccccc1)Cc2ccccc2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?