missing translation for 'onlineSavingsMsg'
Learn More

Dl-2,3-Diaminopropionic Acid, Hydrochloride, TRC

Product Code. 30353539
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30353539 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30353539 Supplier Toronto Research Chemicals Supplier No. TRCD4175751G
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dl-2,3-Diaminopropionic Acid, Hydrochloride
CAS 54897-59-5
Molecular Formula C3H8N2O2 . HCl
Quantity 1 g
Synonym 2,3-Diaminopropanoic Acid Hydrochloride, 2,3-Diaminopropanoic Acid Monohydrochloride, 2,3-Diaminopropionic Acid Hydrochloride, 2,3-Diaminopropionic Acid Monohydrochloride, DL-2,3-Diaminopropionic Acid Hydrochloride, DL-2,3-Diaminopropionic Acid Monohydrochloride, NSC 90287, alpha,beta-Diaminopropionic Acid Monohydrochloride
InChI Formula InChI=1S/C3H8N2O2.ClH/c4-1-2(5)3(6)7;/h2H,1,4-5H2,(H,6,7);1H
SMILES Cl.NCC(N)C(=O)O
IUPAC Name 2,3-diaminopropanoic acid;hydrochloride
Molecular Weight (g/mol) 140.57
Formula Weight 140.0353
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.