missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylglycine, MedChemExpress

Product Code. 16492698
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492698 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16492698 Supplier Medchem Express Supplier No. HYY0069

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress N-Acetylglycine (Aceturic acid) is a minor constituent of numerous foods with no genotoxicity or acute toxicity. N-acetylglycine is used in biological research of peptidomimetics.

N-Acetylglycine (Aceturic acid) is a minor constituent of numerous foods with no genotoxicity or acute toxicity. N-acetylglycine is used in biological research of peptidomimetics.

Specifications

Chemical Name or Material N-Acetylglycine
CAS 543-24-8
Color White
Purity Grade Notes Research
Molecular Formula C4H7NO3
Synonym Aceturic acid Acetamidoacetic acid
Solubility Information DMSO : 100 mg/mL (853.97 mM; Need ultrasonic) ∣H2O : 50 mg/mL (426.99 mM; Need ultrasonic)
SMILES O=C(O)CNC(C)=O
Molecular Weight (g/mol) 117.1
Quantity 5 g
Formula Weight 117.1
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.