missing translation for 'onlineSavingsMsg'
Learn More

6-Morpholinopyridin-3-amine, 97%, Maybridge

Product Code. 10180972
Change view
Click to view available options
:
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10180972 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10180972 Supplier Thermo Scientific Maybridge Supplier No. CD04400DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 52023-68-4
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H13N3O
MDL Number MFCD00119562
Quantity 1 g
Synonym 6-morpholinopyridin-3-amine, 3-amino-6-morpholinopyridine, 6-morpholino-3-pyridinamine, 6-morpholin-4-yl pyridin-3-amine, 6-morpholino pyridin-3-amine, 6-morpholinopyridine-3-amine, 3-pyridinamine, 6-4-morpholinyl, 6-morpholin-4-yl-3-pyridylamine, 5-amino-2-morpholin-4-ylpyridine, 5-amino-2-4-morpholinyl pyridine
InChI Key VVTSPTCBHTWXMD-UHFFFAOYSA-N
SMILES C1COCCN1C2=NC=C(C=C2)N
IUPAC Name 6-morpholin-4-ylpyridin-3-amine
Molecular Weight (g/mol) 179.223
PubChem CID 104059
Formula Weight 179.22
Percent Purity 97%
Chemical Name or Material 6-Morpholinopyridin-3-amine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.