accessibility menu, dialog, popup

UserName

CAS RN 5197-28-4

H3COBrNOO

CAS RN 5197-28-4

IUPAC Name: 2-bromo-1-methoxy-4-nitrobenzene
Synonyms: 2-bromo-1-methoxy-4-nitrobenzene
Molecular Weight (g/mol): 232.03
Molecular Formula: C7H6BrNO3
InChi Key: JMUDXQVNBZCQRF-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1Br)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results