missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4-nitroanisole, 98%, Thermo Scientific™

Product Code. 11377166
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11377166

Brand: Thermo Scientific Alfa Aesar L07181.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5197-28-4
Molecular Formula C7H6BrNO3
Molecular Weight (g/mol) 232.03
MDL Number MFCD00041242
InChI Key JMUDXQVNBZCQRF-UHFFFAOYSA-N
Synonym 2-bromo-4-nitroanisole, 3-bromo-4-methoxynitrobenzene, anisole, 2-bromo-4-nitro, benzene, 2-bromo-1-methoxy-4-nitro, 2-bromo-1-methoxy-4-nitro-benzene, pubchem4133, 2-bromo4-nitroanisole, 4-nitro-2-bromanisol, acmc-1awkm, 2-bromo-4-nitro anisole
PubChem CID 78870
IUPAC Name 2-bromo-1-methoxy-4-nitrobenzene
SMILES COC1=CC=C(C=C1Br)[N+]([O-])=O

Specifications

Melting Point 103°C to 108°C
Quantity 25 g
Beilstein 2556372
Formula Weight 232.03
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-nitroanisole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.