CAS RN 50978-11-5
CAS RN 50978-11-5
IUPAC Name:
3,5-diacetamido-2,4,6-triiodobenzoic acid dihydrate
Synonyms:
amidotrizoic acid dihydrate
Molecular Weight (g/mol):
649.95
Molecular Formula:
C11H13I3N2O6
InChi Key:
JHQKUXXJPHSPOL-UHFFFAOYSA-N
SMILES:
O.O.CC(=O)NC1=C(I)C(C(O)=O)=C(I)C(NC(C)=O)=C1I
1
–
2
of
2
results
2
Amidotrizoic Acid Dihydrate, TRC
CAS: 50978-11-5 Molecular Formula: C11 H9 I3 N2 O4 . 2 H2 O Molecular Weight (g/mol): 649.94 Synonym: Amidotrizoic acid dihydrate,3,5-Bis(acetylamino)-2,4,6-triiodobenzoic acid dihydrate IUPAC Name: 3,5-diacetamido-2,4,6-triiodobenzoic acid;dihydrate SMILES: O.O.CC(=O)Nc1c(I)c(NC(=O)C)c(I)c(C(=O)O)c1I