missing translation for 'onlineSavingsMsg'
Learn More

Diatrizoic acid dihydrate, 98.87%, MP Biomedicals™

Product Code. 11494732 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494732 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11494732 Supplier MP Biomedicals Supplier No. 0219150110
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 50978-11-5
Color White
Assay Percent Range 0.9887
Molecular Formula C11H13I3N2O6
Quantity 10 g
Synonym diatrizoic acid dihydrate, amidotrizoic acid dihydrate, 3,5-diacetamido-2,4,6-triiodobenzoic acid dihydrate, dsstox_cid_28886, dsstox_rid_83154, dsstox_gsid_48960, 2,4,6-triiodo-3,5-diacetamidobenzoic acid dihydrate, 3,5-bis acetylamino-2,4,6-triiodobenzoic acid dihydrate, 3,5-diacetamido-2,4,6-triiodobenzoic acid-water 1/2, amidotrizoic acid ban:jan
InChI Key JHQKUXXJPHSPOL-UHFFFAOYSA-N
SMILES CC(=O)NC1=C(C(=C(C(=C1I)C(=O)O)I)NC(=O)C)I.O.O
IUPAC Name 3,5-diacetamido-2,4,6-triiodobenzoic acid;dihydrate
Molecular Weight (g/mol) 649.946
PubChem CID 12598086
ChEBI CHEBI:59733
Formula Weight 649.95
Physical Form Powder
Chemical Name or Material Diatrizoic acid dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.