accessibility menu, dialog, popup

UserName

NaNaBrBrBrBrPd

CAS RN 50495-13-1

IUPAC Name: palladium(2+) disodium tetrabromide
Synonyms: palladium(2+) disodium tetrabromide
Molecular Weight (g/mol): 472.02
Molecular Formula: Br4Na2Pd
InChi Key: MPLXAXBUDPGDHC-UHFFFAOYSA-J
SMILES: [Na+].[Na+].[Br-].[Br-].[Br-].[Br-].[Pd++]
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results