missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetrabromopalladate(II), Premion™, 99.95% (metals basis), Pd 22.0% min

Product Code. 11324008
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11324008 5 g 5g
11314008 1 g 1g
11334008 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11324008 Supplier Thermo Scientific Alfa Aesar Supplier No. 040407.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50495-13-1 | Br4Na2Pd | 472.016 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50495-13-1
Molecular Formula Br4Na2Pd
Molecular Weight (g/mol) 472.016
MDL Number MFCD00046199
InChI Key MPLXAXBUDPGDHC-UHFFFAOYSA-J
Synonym sodium tetrabromopalladate ii, br4pd.2na, disodium tetrabromopalladiumdiuide, sodium tetrabromopalladate ii , premion
PubChem CID 16217381
IUPAC Name disodium;tetrabromopalladium(2-)
SMILES [Na+].[Na+].Br[Pd-2](Br)(Br)Br

Specifications

Physical Form Powder
Quantity 5 g
Formula Weight 472.02
Percent Purity 99.95%
Grade Premion
Concentration or Composition (by Analyte or Components) Pd 22.0% min.
Assay (metals basis)
Chemical Name or Material Sodium tetrabromopalladate(II)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.