accessibility menu, dialog, popup

UserName

CAS RN 5000-44-2

CH3OSOO

CAS RN 5000-44-2

IUPAC Name: 1-(benzenesulfonyl)propan-2-one
Synonyms: 1-(benzenesulfonyl)propan-2-one
Molecular Weight (g/mol): 198.24
Molecular Formula: C9H10O3S
InChi Key: YBLGSNMIIPIRFC-UHFFFAOYSA-N
SMILES: CC(=O)CS(=O)(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results