CAS RN 499-76-3
CAS RN 499-76-3
IUPAC Name:
4-hydroxy-3-methylbenzoic acid
Synonyms:
4-hydroxy-3-methylbenzoic acid
Molecular Weight (g/mol):
152.15
Molecular Formula:
C8H8O3
InChi Key:
LTFHNKUKQYVHDX-UHFFFAOYSA-N
SMILES:
CC1=CC(=CC=C1O)C(O)=O
1
–
1
of
1
results
1
4-Hydroxy-3-methylbenzoic acid hydrate, 97%, Thermo Scientific™
CAS: 499-76-3 Molecular Formula: C8H8O3 Molecular Weight (g/mol): 152.15 MDL Number: MFCD00270105 InChI Key: LTFHNKUKQYVHDX-UHFFFAOYSA-N Synonym: 4-hydroxy-m-toluic acid,3-methyl-4-hydroxybenzoic acid,4-hydroxy-3-methyl-benzoic acid,4,3-cresotic acid,4-hydroxy-3-methylbenzoate,benzoic acid, 4-hydroxy-3-methyl,4-hydroxy-m-toluate,pubchem20044,acmc-209kj4,3-methyl-4-hydroxybenzoate PubChem CID: 68138 ChEBI: CHEBI:85239 IUPAC Name: 4-hydroxy-3-methylbenzoic acid SMILES: CC1=CC(=CC=C1O)C(O)=O