CAS RN 49759-61-7
CAS RN 49759-61-7
IUPAC Name:
2-amino-3-(4-methylphenyl)propanoic acid
Synonyms:
2-amino-3-(4-methylphenyl)propanoic acid
Molecular Weight (g/mol):
179.22
Molecular Formula:
C10H13NO2
InChi Key:
DQLHSFUMICQIMB-UHFFFAOYNA-N
SMILES:
CC1=CC=C(CC(N)C(O)=O)C=C1
1
–
1
of
1
results
1
4-Methyl-D-phenylalanine, 98%, Thermo Scientific™
CAS: 49759-61-7 Molecular Formula: C10H13NO2 Molecular Weight (g/mol): 179.22 MDL Number: MFCD00152137 InChI Key: DQLHSFUMICQIMB-UHFFFAOYNA-N Synonym: 4-methyl-d-phenylalanine,h-d-phe 4-me-oh,r-2-amino-3-p-tolyl propanoic acid,d-4-methylphenylalanine,d-4-me-phe-oh,2r-2-amino-3-4-methylphenyl propanoic acid,d-phenylalanine, 4-methyl,h-p-me-phe-oh,4-methylphenyl-d-alanine,d-4-methylphe PubChem CID: 7015862 IUPAC Name: (2R)-2-amino-3-(4-methylphenyl)propanoic acid SMILES: CC1=CC=C(CC(N)C(O)=O)C=C1