CAS RN 4936-47-4
CAS RN 4936-47-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Nifuratel, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Nifuratel, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
3
Nifuratel, MedChemExpress
MedChemExpress Nifuratel(NF 113, SAP 113) is a broad antibacterial spectrum agent, which is used as an antibacterial, antifungal, and antiprotozoal (Trichomonas).