missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nifuratel, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Nifuratel
Specifications
Specifications
| Chemical Name or Material | Nifuratel |
| CAS | 4936-47-4 |
| Analyte or Component Names | Nifuratel |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H11 N3 O5 S |
| Certifications/Compliance | ISO 17034 |
| Synonym | 2-Oxazolidinone, 5-[(methylthio)methyl]-3-[[(5-nitro-2-furanyl)methylene]amino]-, 2-Oxazolidinone, 5-[(methylthio)methyl]-3-[(5-nitrofurfurylidene)amino]- (7CI,8CI), 3-[(5-Nitro-2-furfurylidene)amino]-5-[(methylthio)methyl]-2-oxazolidinone, 5-(Methylmercaptomethyl)-3-(5-nitro-2-furfurylidenamino)-2-oxazolidinone, 5-(Methylthiomethyl)-3-(5-nitro-2-furfurylidenamino)oxazolidin-2-one, 5-[(Methylthio)methyl]-3-[(5-nitrofurfurylidene)amino]-2-oxazolidinone, Inimur, Macmiror, Magmilor, Methylmercadone, NF 113, Nifuratel, Omnes, Polmiror, SAP 113, Tydantil |
| InChI Formula | InChI=1S/C10H11N3O5S/c1-19-6-8-5-12(10(14)18-8)11-4-7-2-3-9(17-7)13(15)16/h2-4,8H,5-6H2,1H3/b11-4+ |
| SMILES | CSCC1CN(\N=C\c2oc(cc2)[N+](=O)[O-])C(=O)O1 |
| IUPAC Name | 5-(methylsulfanylmethyl)-3-[(E)-(5-nitrofuran-2-yl)methylideneamino]-1,3-oxazolidin-2-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction