missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitroso-1-piperazineethanol, Mikromol
Description
4-Nitroso-1-piperazineethanol, Mikromol
Specifications
Specifications
| Chemical Name or Material | Aripiprazole Impurity 30 |
| CAS | 48121-20-6 |
| Analyte or Component Names | Aripiprazole Impurity 30 |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H13 N3 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 4-Nitroso-1-piperazineethanol, 2-(4-Nitroso-piperazin-1-yl)-ethanol, 2-(4-Nitrosopiperazin-1-yl)ethan-1-ol, 4-Nitrosopiperazine-1-ethanol |
| InChI Formula | InChI=1S/C6H13N3O2/c10-6-5-8-1-3-9(7-11)4-2-8/h10H,1-6H2 |
| Solution Type | Neat |
| SMILES | OCCN1CCN(CC1)N=O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?