accessibility menu, dialog, popup

UserName

CAS RN 4779-31-1

OHOHNOOOO

CAS RN 4779-31-1

IUPAC Name: 4-(benzyloxy)-3-{[(benzyloxy)carbonyl]amino}-4-oxobutanoic acid
Synonyms: 4-(benzyloxy)-3-{[(benzyloxy)carbonyl]amino}-4-oxobutanoic acid
Molecular Weight (g/mol): 357.36
Molecular Formula: C19H19NO6
InChi Key: UYOZWZKJQRBZRH-UHFFFAOYNA-N
SMILES: OC(=O)CC(NC(=O)OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)