missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbobenzyloxy-L-aspartic Acid 1-Benzyl Ester, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | N-Carbobenzyloxy-L-aspartic Acid 1-Benzyl Ester |
| CAS | 4779-31-1 |
| Molecular Formula | C19 H19 N O6 |
| Quantity | 1 g |
| Synonym | L-Aspartic acid, N-[(phenylmethoxy)carbonyl]-, 1-(phenylmethyl) ester, Aspartic acid, N-carboxy-, N,1-dibenzyl ester (7CI), Aspartic acid, N-carboxy-, N,1-dibenzyl ester, L- (8CI), 1-Benzyl N-benzyloxycarbonyl-L-aspartate, N-(Benzyloxycarbonyl)-L-aspartic acid benzyl ester, N-(Benzyloxycarbonyl)aspartic acid α-benzyl ester, N-(α-Carbobenzoxy)-L-aspartic acid α-benzyl ester, NSC 150012, NSC 173727, α-Benzyl N-(benzyloxycarbonyl)-L-aspartate, Z-Asp-OBzl, 1-Benzyl N-carbobenzyloxy-L-aspartate, Cbz-L-Asp-OBzl, (3S)-4-(Benzyloxy)-3-{[(benzyloxy)carbonyl]amino}-4-oxobutanoic acid, N-Benzyloxycarbonyl-L-aspartic acid 1-benzyl ester |
| InChI Formula | InChI=1S/C19H19NO6/c21-17(22)11-16(18(23)25-12-14-7-3-1-4-8-14)20-19(24)26-13-15-9-5-2-6-10-15/h1-10,16H,11-13H2,(H,20,24)(H,21,22)/t16-/m0/s1 |
| SMILES | OC(=O)C[C@H](NC(=O)OCc1ccccc1)C(=O)OCc2ccccc2 |
| IUPAC Name | (3S)-4-oxo-4-phenylmethoxy-3-(phenylmethoxycarbonylamino)butanoic acid |
| Molecular Weight (g/mol) | 357.36 |
| Formula Weight | 357.1212 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction