missing translation for 'onlineSavingsMsg'
Learn More

N-Carbobenzyloxy-L-aspartic Acid 1-Benzyl Ester, TRC

Product Code. 30350530
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 30350530

Brand: Toronto Research Chemicals TRCC1763005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Carbobenzyloxy-L-aspartic Acid 1-Benzyl Ester
CAS 4779-31-1
Molecular Formula C19 H19 N O6
Quantity 5 g
Synonym L-Aspartic acid, N-[(phenylmethoxy)carbonyl]-, 1-(phenylmethyl) ester, Aspartic acid, N-carboxy-, N,1-dibenzyl ester (7CI), Aspartic acid, N-carboxy-, N,1-dibenzyl ester, L- (8CI), 1-Benzyl N-benzyloxycarbonyl-L-aspartate, N-(Benzyloxycarbonyl)-L-aspartic acid benzyl ester, N-(Benzyloxycarbonyl)aspartic acid α-benzyl ester, N-(α-Carbobenzoxy)-L-aspartic acid α-benzyl ester, NSC 150012, NSC 173727, α-Benzyl N-(benzyloxycarbonyl)-L-aspartate, Z-Asp-OBzl, 1-Benzyl N-carbobenzyloxy-L-aspartate, Cbz-L-Asp-OBzl, (3S)-4-(Benzyloxy)-3-{[(benzyloxy)carbonyl]amino}-4-oxobutanoic acid, N-Benzyloxycarbonyl-L-aspartic acid 1-benzyl ester
InChI Formula InChI=1S/C19H19NO6/c21-17(22)11-16(18(23)25-12-14-7-3-1-4-8-14)20-19(24)26-13-15-9-5-2-6-10-15/h1-10,16H,11-13H2,(H,20,24)(H,21,22)/t16-/m0/s1
SMILES OC(=O)C[C@H](NC(=O)OCc1ccccc1)C(=O)OCc2ccccc2
IUPAC Name (3S)-4-oxo-4-phenylmethoxy-3-(phenylmethoxycarbonylamino)butanoic acid
Molecular Weight (g/mol) 357.36
Formula Weight 357.1212
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.