CAS RN 461000-66-8
CAS RN 461000-66-8
IUPAC Name:
3,4,5-trimethoxy-N-(2-methoxyethyl)-N-(4-phenyl-1,3-thiazol-2-yl)benzamide
Synonyms:
3,4,5-trimethoxy-N-(2-methoxyethyl)-N-(4-phenyl-1,3-thiazol-2-yl)benzamide
Molecular Weight (g/mol):
428.5
Molecular Formula:
C22H24N2O5S
InChi Key:
ZUXNHFFVQWADJL-UHFFFAOYSA-N
SMILES:
COCCN(C(=O)C1=CC(OC)=C(OC)C(OC)=C1)C1=NC(=CS1)C1=CC=CC=C1