CAS RN 4513-62-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,6-dimethyl 2-methyl-5-methylidenehexanedioate, TRC
CAS: 4513-62-6 Molecular Formula: C10H16O4 Molecular Weight (g/mol): 200.23 IUPAC Name: dimethyl 2-methyl-5-methylidenehexanedioate SMILES: CC(CCC(=C)C(=O)OC)C(=O)OC