missing translation for 'onlineSavingsMsg'
Learn More

1,6-dimethyl 2-methyl-5-methylidenehexanedioate, TRC

Product Code. 30387998
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387998 50 mg
30382135 10 mg
30371192 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387998 Supplier Toronto Research Chemicals Supplier No. TRCD48257850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4513-62-6
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.23
IUPAC Name dimethyl 2-methyl-5-methylidenehexanedioate
SMILES CC(CCC(=C)C(=O)OC)C(=O)OC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H16O4/c1-7(9(11)13-3)5-6-8(2)10(12)14-4/h8H,1,5-6H2,2-4H3
Formula Weight 200.1049
Chemical Name or Material 1,6-dimethyl 2-methyl-5-methylidenehexanedioate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.