accessibility menu, dialog, popup

UserName

CAS RN 4461-29-4

NH2OS

CAS RN 4461-29-4

IUPAC Name: 2-(thiophen-2-yl)acetamide
Synonyms: thiophen-2-yl-acetamide
Molecular Weight (g/mol): 141.19
Molecular Formula: C6H7NOS
InChi Key: UUPZTFTUZUQRQT-UHFFFAOYSA-N
SMILES: NC(=O)CC1=CC=CS1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)