missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Thiophen-2-yl)acetamide, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 2-(Thiophen-2-yl)acetamide |
| CAS | 4461-29-4 |
| Molecular Formula | C6H7NOS |
| Quantity | 100 mg |
| Synonym | 2-Thienylacetamide |
| InChI Formula | InChI=1S/C6H7NOS/c7-6(8)4-5-2-1-3-9-5/h1-3H,4H2,(H2,7,8) |
| SMILES | NC(=O)Cc1cccs1 |
| IUPAC Name | 2-thiophen-2-ylacetamide |
| Molecular Weight (g/mol) | 141.19 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction