accessibility menu, dialog, popup

UserName

CAS RN 446053-24-3

NHOFFFCl

CAS RN 446053-24-3

IUPAC Name: N-(2-chlorophenyl)-4-(trifluoromethyl)benzamide
Synonyms: N-(2-chlorophenyl)-4-(trifluoromethyl)benzamide
Molecular Weight (g/mol): 299.68
Molecular Formula: C14H9ClF3NO
InChi Key: DFDOVSPUQBTDBB-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(C=C1)C(=O)NC1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results