accessibility menu, dialog, popup

UserName

CAS RN 446-07-1

CH3FH3CO

CAS RN 446-07-1

IUPAC Name: 1-(2-fluoro-5-methylphenyl)ethan-1-one
Synonyms: 1-(2-fluoro-5-methylphenyl)ethanone
Molecular Weight (g/mol): 152.17
Molecular Formula: C9H9FO
InChi Key: DNDPUIGGUBKRQB-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(C)=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results