accessibility menu, dialog, popup

UserName

HClH3COClH2N

CAS RN 4274-03-7

IUPAC Name: hydrogen 5-chloro-2-methoxyaniline chloride
Synonyms: hydrogen 5-chloro-2-methoxyaniline chloride
Molecular Weight (g/mol): 194.06
Molecular Formula: C7H9Cl2NO
InChi Key: ASEYMHNCCXSHIL-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC1=CC=C(Cl)C=C1N
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results