missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methoxyaniline hydrochloride, 98%, Thermo Scientific™

Product Code. 11445333
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445333 500 g 500g
11435333 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445333 Supplier Thermo Scientific Alfa Aesar Supplier No. A12887.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4274-03-7
MDL Number MFCD00035488

Specifications

Melting Point ∼228°C (decomposition)
Molecular Formula C5H13NO
Quantity 500 g
UN Number UN2811
Synonym d-valinol, r---2-amino-3-methyl-1-butanol, r-2-amino-3-methylbutan-1-ol, 2r-2-amino-3-methylbutan-1-ol, r-2-amino-3-methyl-1-butanol, d-2-amino-3-methyl-1-butanol, 2r-2-amino-3-methyl-1-butanol, 1r-1-isopropyl-2-hydroxyethylamine, d---valinol r---2-amino-3-methyl-1-butanol, d-valinol r---2-amino-3-methyl-1-butanol
InChI Key NWYYWIJOWOLJNR-YFKPBYRVSA-N
SMILES CC(C)[C@@H](N)CO
IUPAC Name (2R)-2-amino-3-methylbutan-1-ol
Molecular Weight (g/mol) 103.17
PubChem CID 6950587
Formula Weight 194.06
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-methoxyaniline hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.