missing translation for 'onlineSavingsMsg'
Learn More

Dichloroacetic Anhydride (~85%), TRC

Product Code. 30365611
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30365611 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30365611 Supplier Toronto Research Chemicals Supplier No. TRCD1897601G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dichloroacetic anhydride
CAS 4124-30-5
Molecular Formula C4 H2 Cl4 O3
Quantity 1 g
InChI Formula InChI=1S/C4H2Cl4O3/c5-1(6)3(9)11-4(10)2(7)8/h1-2H
SMILES ClC(Cl)C(=O)OC(=O)C(Cl)Cl
IUPAC Name (2,2-dichloroacetyl) 2,2-dichloroacetate
Molecular Weight (g/mol) 239.869
Formula Weight 237.876
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.