accessibility menu, dialog, popup

UserName

CAS RN 407-41-0

NH2(S)OPHOHOOOHO

CAS RN 407-41-0

IUPAC Name: (2S)-2-amino-3-(phosphonooxy)propanoic acid
Synonyms: phosphoserine phosphonoserine dexfosfoserine
Molecular Weight (g/mol): 185.07
Molecular Formula: C3H8NO6P
InChi Key: BZQFBWGGLXLEPQ-REOHCLBHSA-N
SMILES: N[C@@H](COP(O)(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results