missing translation for 'onlineSavingsMsg'
Learn More

O-Phospho-L-serine, MedChemExpress

Product Code. 16486636
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16486636

Brand: Medchem Express HY15129

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress O-Phospho-L-serine is the immediate precursor to L-serine in the serine synthesis pathway, and an agonist at the group III mGluR receptors (mGluR4, mGluR6, mGluR7, and mGluR8); O-Phospho-L-serine also acts as a weak antagonist for mGluR1 and a potent antagonist for mGluR2.

O-Phospho-L-serine is the immediate precursor to L-serine in the serine synthesis pathway, and an agonist at the group III mGluR receptors (mGluR4, mGluR6, mGluR7, and mGluR8)

Specifications

Chemical Name or Material O-Phospho-L-serine
Physical Form Solid
CAS 407-41-0
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C3H8NO6P
For Use With (Application) Neuroscience-Neuromodulation
Synonym L-Serine O-phosphate L-SOP
Solubility Information H2O : < 0.1 mg/mL (insoluble) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES OP(OC[C@@H](C(O)=O)N)(O)=O
Molecular Weight (g/mol) 185.07
Formula Weight 185.07
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.