accessibility menu, dialog, popup

UserName

CAS RN 40674-21-3

H3COOCH3H3CONOO

CAS RN 40674-21-3

IUPAC Name: ethyl 2-[(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)oxy]-2-methylpropanoate
Synonyms: ethyl 2-[(1,3-dioxoisoindol-2-yl)oxy]-2-methylpropanoate
Molecular Weight (g/mol): 277.28
Molecular Formula: C14H15NO5
InChi Key: UNZUMKCIIIFEJI-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C)(C)ON1C(=O)C2=CC=CC=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results