accessibility menu, dialog, popup

UserName

CAS RN 405554-55-4

CH3NH3CONNOHNNH(E)

CAS RN 405554-55-4

IUPAC Name: (2-{4-[5-(1,4-dihydropyridin-4-ylidene)-4-(3-nitroso-2,6-dihydro-1H-inden-6-ylidene)-4,5-dihydro-1H-imidazol-2-yl]phenoxy}ethyl)dimethylamine
Synonyms: dimethyl(2-{4-[4-(1-nitroso-2,3-dihydroinden-5-ylidene)-5-(1H-pyridin-4-ylidene)-1H-imidazol-2-yl]phenoxy}ethyl)amine
Molecular Weight (g/mol): 453.55
Molecular Formula: C27H27N5O2
InChi Key: DPCXEUSDRQOOGZ-UHFFFAOYSA-N
SMILES: CN(C)CCOC1=CC=C(C=C1)C1=NC(C(N1)=C1C=CNC=C1)=C1C=CC2=C(CCC2=C1)N=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results