accessibility menu, dialog, popup

UserName

CAS RN 402-10-8

H3COFFFBr

CAS RN 402-10-8

IUPAC Name: 2-bromo-1-methoxy-4-(trifluoromethyl)benzene
Synonyms: 2-bromo-1-methoxy-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 255.03
Molecular Formula: C8H6BrF3O
InChi Key: OWUAHASLKZBQTD-UHFFFAOYSA-N
SMILES: COC1=C(Br)C=C(C=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results