accessibility menu, dialog, popup

UserName

CAS RN 401575-24-4

NHNOFFF

CAS RN 401575-24-4

IUPAC Name: 2,2,2-trifluoro-N-[(pyridin-2-yl)methyl]acetamide
Synonyms: 2,2,2-trifluoro-N-(pyridin-2-ylmethyl)acetamide
Molecular Weight (g/mol): 204.15
Molecular Formula: C8H7F3N2O
InChi Key: SCEMKHWGDPXKIJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)NCC1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results