accessibility menu, dialog, popup

UserName

H2OH2OH2OH2OOHNiOOO

CAS RN 39430-27-8

IUPAC Name: nickel(2+) tetrahydrate hydroxide carbonate
Synonyms: nickel(2+) tetrahydrate hydroxide carbonate
Molecular Weight (g/mol): 207.77
Molecular Formula: CH9NiO8
InChi Key: KNXPLWTUEQLSEP-UHFFFAOYSA-K
SMILES: O.O.O.O.[OH-].[Ni++].[O-]C([O-])=O
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results