missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) carbonate hydroxide hydrate, for analysis, (ca 45-47% Ni)

Product Code. 10370644 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10370644 250 g Plastic bottle 250g
10360644 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10370644 Supplier Thermo Scientific Acros Supplier No. 223162500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 39430-27-8 | CH4Ni3O7·xH2O | 304.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39430-27-8 , 65405-96-1
Molecular Formula CH4Ni3O7·xH2O
Molecular Weight (g/mol) 304.11
MDL Number MFCD00150262
Synonym Basic nickel carbonate

Specifications

Color Green
Physical Form Powder
Quantity 250 g
Linear Formula 2Ni(OH)2·xH2O
Merck Index 13, 6527
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ammonia and in diluted acids
Formula Weight 304.11
Percent Purity ≥45% (NI)
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Nickel(II) carbonate hydroxide hydrate, ca 45-47% Ni, For analysis
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.