accessibility menu, dialog, popup

UserName

SOOClFFFNOO

CAS RN 39234-83-8

IUPAC Name: 4-nitro-3-(trifluoromethyl)benzene-1-sulfonyl chloride
Synonyms: 4-nitro-3-(trifluoromethyl)benzenesulfonyl chloride
Molecular Weight (g/mol): 289.61
Molecular Formula: C7H3ClF3NO4S
InChi Key: LJQUNSRHHHYXEW-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(C=C(C=C1)S(Cl)(=O)=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results