missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-3-(trifluoromethyl)benzenesulfonyl chloride, 97%

Product Code. 11375555
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11375555 5 g 5g
11365555 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11375555 Supplier Thermo Scientific Alfa Aesar Supplier No. L19721.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 39234-83-8 | C7H3ClF3NO4S | 289.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39234-83-8
Molecular Formula C7H3ClF3NO4S
Molecular Weight (g/mol) 289.61
MDL Number MFCD03094387
InChI Key LJQUNSRHHHYXEW-UHFFFAOYSA-N
Synonym 4-nitro-3-trifluoromethyl benzenesulfonyl chloride, 4-nitro-3-trifluoromethyl benzenesulphonyl chloride, 4-nitro-3-trifluoromethyl benzenesulfonylchloride, 4-nitro-3-trifluoromethyl benzene-1-sulfonyl chloride, pubchem11722, acmc-1ail1, 5-chlorosulphonyl-2-nitrobenzotrifluoride, 4-nitro-3-trifluoromethyl phenylsulfonyl chloride, 4-chlorosulphonyl-2-trifluoromethyl nitrobenzene, 4-nitro-3-trifluoromethyl-benzenesulfonyl chloride
PubChem CID 2778595
IUPAC Name 4-nitro-3-(trifluoromethyl)benzenesulfonyl chloride
SMILES [O-][N+](=O)C1=C(C=C(C=C1)S(Cl)(=O)=O)C(F)(F)F

Specifications

Melting Point 63°C to 65°C
Quantity 5 g
UN Number UN3261
Beilstein 2880842
Sensitivity Moisture sensitive
Formula Weight 289.62
Percent Purity 97%
Chemical Name or Material 4-Nitro-3-(trifluoromethyl)benzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.