CAS RN 38862-65-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ethyl Olivetolate, TRC
CAS: 38862-65-6 Molecular Formula: C14H20O4 Molecular Weight (g/mol): 252.31 Synonym: Ethyl 2,4-Dihydroxy-6-pentylbenzoate SMILES: CCCCCc1cc(O)cc(O)c1C(=O)OCC