missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Olivetolate, TRC

Product Code. 30381122
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381122 5 g -
30377161 1 g -
30379445 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381122 Supplier Toronto Research Chemicals Supplier No. TRCE3886205G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38862-65-6
Molecular Formula C14H20O4
Molecular Weight (g/mol) 252.31
Synonym Ethyl 2,4-Dihydroxy-6-pentylbenzoate
SMILES CCCCCc1cc(O)cc(O)c1C(=O)OCC

Specifications

Quantity 5 g
InChI Formula InChI=1S/C14H20O4/c1-3-5-6-7-10-8-11(15)9-12(16)13(10)14(17)18-4-2/h8-9,15-16H,3-7H2,1-2H3
Formula Weight 252.1362
Chemical Name or Material Ethyl Olivetolate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.