missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-thiopheneacetate, 98%, ACROS Organics™

Product Code. 10236580 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
Packaging:
Glass bottle
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10236580 10 mL Glass bottle 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10236580 Supplier Thermo Scientific Acros Supplier No. 241910100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 37784-63-7
Color Colorless to Brown
Density 1.1200g/mL
Boiling Point 97.0°C to 98.0°C (8.0 mmHg)
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C8H10O2S
Refractive Index 1.5090 to 1.5110
MDL Number MFCD00005472
Quantity 10 mL
Beilstein 18, III, 4066
Specific Gravity 1.12
Synonym ethyl thiophene-3-acetate, ethyl 3-thienylacetate, ethyl 3-thiopheneacetate, ethyl 2-thiophen-3-yl acetate, 3-thiopheneacetic acid, ethyl ester, ethyl 3-thienylacetate #, acmc-1afw1, ethyl 2-3-thienyl acetate, bidd:gt0101, ethyl 2-thiophen-3-ylethanoate
InChI Key FZBNQIWPYCUPAP-UHFFFAOYSA-N
SMILES CCOC(=O)CC1=CSC=C1
IUPAC Name ethyl 2-(thiophen-3-yl)acetate
Molecular Weight (g/mol) 170.23
PubChem CID 520865
Formula Weight 170.23
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 3-thiopheneacetate, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.