CAS RN 375345-95-2
CAS RN 375345-95-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Mibampator, MedChemExpress
MedChemExpress Mibampator (LY451395) is a potent and highly selective potentiator of the AMPA receptors.