missing translation for 'onlineSavingsMsg'
Learn More

Mibampator, MedChemExpress

Product Code. 16465216
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465216 100 mg 100mg
16427017 5 mg 5mg
16420627 50 mg 50mg
16414298 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16465216 Supplier Medchem Express Supplier No. HY10934
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Mibampator (LY451395) is a potent and highly selective potentiator of the AMPA receptors.

Mibampator (LY451395) is a potent and highly selective potentiator of the AMPA receptors.

Specifications

Chemical Name or Material Mibampator
CAS 375345-95-2
Color White
Purity Grade Notes Research
Molecular Formula C21H30N2O4S2
Quantity 100 mg
Synonym LY451395
Solubility Information DMSO : ≥ 25 mg/mL (57.00 mM)
SMILES C[C@@H](CNS(C(C)C)(=O)=O)C1=CC=C(C2=CC=C(CCNS(C)(=O)=O)C=C2)C=C1
Molecular Weight (g/mol) 438.6
Formula Weight 438.6
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.