CAS RN 374564-35-9
IUPAC Name:
potassium (4-bromophenyl)trifluoroboranuide
Synonyms:
potassium (4-bromophenyl)trifluoroboranuide
Molecular Weight (g/mol):
262.91
Molecular Formula:
C6H4BBrF3K
InChi Key:
YTSKPQQWISZEQO-UHFFFAOYSA-N
SMILES:
[K+].F[B-](F)(F)C1=CC=C(Br)C=C1
1
–
1
of
1
results
1
Potassium 4-bromophenyltrifluoroborate, 97%, Thermo Scientific™
CAS: 374564-35-9 Molecular Formula: C6H4BBrF3K Molecular Weight (g/mol): 262.91 MDL Number: MFCD02093978 InChI Key: YTSKPQQWISZEQO-UHFFFAOYSA-N Synonym: potassium 4-bromophenyl trifluoroborate,potassium 4-bromophenyltrifluoroborate,potassium 4-bromophenyl trifluoroboranuide,pubchem11345,amtb079,potassium 4-bromophenyl-trifluoroboranuide,potassium ion 4-bromophenyl trifluoroboranuide,potassium 4-bromophenyl trifluoro borate 1-,potassium 4-bromophenyl-tris fluoranyl boranuide,4-bromophenyltrifluoroboric acid potassium salt PubChem CID: 23674779 SMILES: [K+].F[B-](F)(F)C1=CC=C(Br)C=C1