missing translation for 'onlineSavingsMsg'
Learn More

Potassium 4-bromophenyltrifluoroborate, 97%, Thermo Scientific™

Product Code. 11324855
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11324855 5 g 5g
11314855 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11324855 Supplier Thermo Scientific Alfa Aesar Supplier No. L17967.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Potassium 4-bromophenyltrifluoroborate is used as potent boronic acid surrogates in Suzuki-Miyaura Cross-Coupling reactions.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Moisture sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 374564-35-9
Molecular Formula C6H4BBrF3K
Molecular Weight (g/mol) 262.91
MDL Number MFCD02093978
InChI Key YTSKPQQWISZEQO-UHFFFAOYSA-N
Synonym potassium 4-bromophenyl trifluoroborate, potassium 4-bromophenyltrifluoroborate, potassium 4-bromophenyl trifluoroboranuide, pubchem11345, amtb079, potassium 4-bromophenyl-trifluoroboranuide, potassium ion 4-bromophenyl trifluoroboranuide, potassium 4-bromophenyl trifluoro borate 1-, potassium 4-bromophenyl-tris fluoranyl boranuide, 4-bromophenyltrifluoroboric acid potassium salt
PubChem CID 23674779
SMILES [K+].F[B-](F)(F)C1=CC=C(Br)C=C1

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 9297533
Solubility Information Soluble in water.
IUPAC Name potassium;(4-bromophenyl)-trifluoroboranuide
Formula Weight 262.91
Percent Purity 97%
Chemical Name or Material Potassium 4-bromophenyltrifluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.