accessibility menu, dialog, popup

UserName

CAS RN 36880-33-8

H3CSO

CAS RN 36880-33-8

IUPAC Name: 5-ethylthiophene-2-carbaldehyde
Synonyms: 5-ethylthiophene-2-carbaldehyde
Molecular Weight (g/mol): 140.2
Molecular Formula: C7H8OS
InChi Key: CLQXZICUPGZTPE-UHFFFAOYSA-N
SMILES: CCC1=CC=C(S1)C=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results