CAS RN 361543-99-9
CAS RN 361543-99-9
IUPAC Name:
(4-methoxy-2,6-dimethylphenyl)boronic acid
Synonyms:
4-methoxy-2,6-dimethylphenylboronic acid
Molecular Weight (g/mol):
180.01
Molecular Formula:
C9H13BO3
InChi Key:
UOSSBXMFWPYHEF-UHFFFAOYSA-N
SMILES:
COC1=CC(C)=C(B(O)O)C(C)=C1
1
–
1
of
1
results
1
4-Methoxy-2,6-dimethylbenzeneboronic acid, 95%, Thermo Scientific™
CAS: 361543-99-9 Molecular Formula: C9H13BO3 Molecular Weight (g/mol): 180.01 MDL Number: MFCD03452755 InChI Key: UOSSBXMFWPYHEF-UHFFFAOYSA-N Synonym: 2,6-dimethyl-4-methoxyphenyl boronic acid,2,6-dimethyl-4-methoxyphenylboronic acid,4-methoxy-2,6-dimethylphenyl boronic acid,2,6-dimethyl-4-methoxybenzeneboronic acid,2-borono-5-methoxy-m-xylene,4-borono-3,5-dimethylanisole,4-methoxy-2,6-dimethylbenzeneboronic acid,4-methoxy-2,6-dimethylphenylboronicacid,boronic acid, 4-methoxy-2,6-dimethylphenyl,acmc-209il9 PubChem CID: 3716303 IUPAC Name: (4-methoxy-2,6-dimethylphenyl)boronic acid SMILES: B(C1=C(C=C(C=C1C)OC)C)(O)O